missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diethoxypropionitrile 98.0+%, TCI America™

Catalog No. D28665ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28665ML 5 mL
D286625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28665ML Supplier TCI America Supplier No. D28665ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56011-12-2
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD00209573
InChI Key QPFCLGADWIWYFW-UHFFFAOYSA-N
PubChem CID 566609
IUPAC Name 2,2-diethoxypropanenitrile
SMILES CCOC(C)(C#N)OCC

Specifications

Color Colorless
Boiling Point 79°C
Quantity 5 mL
UN Number 1993
Formula Weight 143.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Diethoxypropionitrile