missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Diethoxypropane 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 113°C |
| Quantity | 250 mL |
| UN Number | 1993 |
| Formula Weight | 132.20 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,2-Diethoxypropane |
Safety and Handling
| RTECSNumber | AL4900000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |