missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diethoxyacetophenone 95.0+%, TCI America™

Catalog No. D164025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D164025G 25 g
D1640500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D164025G Supplier TCI America Supplier No. D164025G

Chemical Identifiers

CAS 6175-45-7
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.26
MDL Number MFCD00009659
InChI Key PIZHFBODNLEQBL-UHFFFAOYSA-N
Synonym 2,2-diethoxyacetophenone, ethanone, 2,2-diethoxy-1-phenyl, phenylglyoxal diethyl acetal, 2,2-diethoxy-1-phenylethan-1-one, alpha,alpha-diethoxyacetophenone, unii-w4bcz1mak3, 2,2-diethoxy acetophenone, ccris 5222, benzoylformaldehyde diethyl acetal, glyoxal, phenyl-, 2-diethyl acetal
PubChem CID 22555
IUPAC Name 2,2-diethoxy-1-phenylethan-1-one
SMILES CCOC(OCC)C(=O)C1=CC=CC=C1

Specifications

Boiling Point 134°C
Quantity 25 g
Formula Weight 208.26
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Diethoxyacetophenone
EINECSNumber (3)-3593
RTECSNumber MD3284000
TSCA Yes