missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dichloropropane 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -35°C |
| Color | Colorless |
| Boiling Point | 69°C |
| Quantity | 10 mL |
| UN Number | 1993 |
| Formula Weight | 112.98 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,2-Dichloropropane |
Safety and Handling
| RTECSNumber | TX9662500 |
| TSCA | Yes |