missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2,2-Dichlorocyclopropyl)benzene 98.0+%, TCI America™
Supplier: TCI America D441025G
Specifications
| (2,2-Dichlorocyclopropyl)benzene | |
| Yellow | |
| C9H8Cl2 | |
| 25 g | |
| WLWFQGXZIDYWQF-UHFFFAOYSA-N | |
| (2,2-dichlorocyclopropyl)benzene | |
| 95349 | |
| ≥98.0% (GC) |
| 2415-80-7 | |
| 105°C | |
| MFCD00001272 | |
| 2,2-dichlorocyclopropyl benzene, 1,1-dichloro-2-phenylcyclopropane, 2,2-dichlorocyclopropyl-benzene, benzene, 2,2-dichlorocyclopropyl, cyclopropane, 1,1-dichloro, 2-phenyl, pubchem18679, acmc-1clka, ksc491m2n, wlwfqgxzidywqf-uhfffaoysa, 2,2-dichloro-cyclopropyl-benzene | |
| C1C(C1(Cl)Cl)C2=CC=CC=C2 | |
| 187.063 | |
| 187.06 | |
| Liquid |
Chemical Identifiers
| 2415-80-7 | |
| 187.063 | |
| WLWFQGXZIDYWQF-UHFFFAOYSA-N | |
| 95349 | |
| C1C(C1(Cl)Cl)C2=CC=CC=C2 |
| C9H8Cl2 | |
| MFCD00001272 | |
| 2,2-dichlorocyclopropyl benzene, 1,1-dichloro-2-phenylcyclopropane, 2,2-dichlorocyclopropyl-benzene, benzene, 2,2-dichlorocyclopropyl, cyclopropane, 1,1-dichloro, 2-phenyl, pubchem18679, acmc-1clka, ksc491m2n, wlwfqgxzidywqf-uhfffaoysa, 2,2-dichloro-cyclopropyl-benzene | |
| (2,2-dichlorocyclopropyl)benzene |
Safety and Handling
TSCA : No