missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dichlorobenzil, 98+%

Catalog No. AAL0650714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0650714 25 g
AAL0650706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0650714 Supplier Thermo Scientific Chemicals Supplier No. L0650714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21854-95-5 | C14H8Cl2O2 | 279.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21854-95-5
Molecular Formula C14H8Cl2O2
Molecular Weight (g/mol) 279.116
MDL Number MFCD00018263
InChI Key VOSNNSVWVJFJCR-UHFFFAOYSA-N
Synonym 2,2'-dichlorobenzil, 1,2-bis 2-chlorophenyl ethane-1,2-dione, ethanedione, bis 2-chlorophenyl, 2,2'-dichlorodibenzoyl, benzil, 2,2'-dichloro, 1,2-ethanedione, 1,2-bis 2-chlorophenyl, bis 2-chlorophenyl ethane-1,2-dione, benzil,2'-dichloro, 2,2/'-dichlorobenzil, acmc-20aok0
PubChem CID 89076
IUPAC Name 1,2-bis(2-chlorophenyl)ethane-1,2-dione
SMILES C1=CC=C(C(=C1)C(=O)C(=O)C2=CC=CC=C2Cl)Cl

Specifications

Melting Point 132°C to 136°C
Quantity 25 g
Beilstein 2120689
Formula Weight 279.12
Percent Purity ≥98%
Chemical Name or Material 2,2'-Dichlorobenzil
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only