missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dichloroacetamide 98.0+%, TCI America™

Catalog No. D030625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D030625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D030625G Supplier TCI America Supplier No. D030625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 683-72-7
Molecular Formula C2H3Cl2NO
Molecular Weight (g/mol) 127.952
MDL Number MFCD00008015
InChI Key WCGGWVOVFQNRRS-UHFFFAOYSA-N
Synonym dichloroacetamide, acetamide, 2,2-dichloro, acetamide, dichloro, unii-i202lta03d, 2,2'-dichloroacetamide, 2,2-dichloro-acetamide, acetamide,2,2-dichloro, acmc-1aum5, dsstox_cid_1561, wln: gngv1
PubChem CID 12694
IUPAC Name 2,2-dichloroacetamide
SMILES C(C(=O)N)(Cl)Cl

Specifications

Melting Point 99°C
Color White
Boiling Point 234°C
Quantity 25 g
Formula Weight 127.95
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Dichloroacetamide
RTECSNumber AB6475000
TSCA Yes