missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dichloroacetamide, 98+%

Catalog No. AAL0031509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0031509 10 g
AAL0031518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0031509 Supplier Thermo Scientific Chemicals Supplier No. L0031509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 683-72-7 | C2H3Cl2NO | 127.952 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 683-72-7
Molecular Formula C2H3Cl2NO
Molecular Weight (g/mol) 127.952
MDL Number MFCD00008015
InChI Key WCGGWVOVFQNRRS-UHFFFAOYSA-N
PubChem CID 12694
IUPAC Name 2,2-dichloroacetamide
SMILES C(C(=O)N)(Cl)Cl

Specifications

Melting Point 96°C to 101°C
Boiling Point 233°C to 234°C
Linear Formula Cl2CHCONH2
Quantity 10 g
Beilstein 1743236
Synonym dichloroacetamide, acetamide, 2,2-dichloro, acetamide, dichloro, unii-i202lta03d, 2,2'-dichloroacetamide, 2,2-dichloro-acetamide, acetamide,2,2-dichloro, acmc-1aum5, dsstox_cid_1561, wln: gngv1
Formula Weight 127.96
Percent Purity ≥98%
Chemical Name or Material 2,2-Dichloroacetamide
Hazard Category H301+H311-H317
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P310-P302+P352-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 211-674-2
RTECSNumber AB6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only