missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dibromo-2-cyanoacetamide 98.0+%, TCI America™
$57.79 - $328.08
Chemical Identifiers
| CAS | 10222-01-2 |
|---|---|
| Molecular Formula | C3H2Br2N2O |
| Molecular Weight (g/mol) | 241.87 |
| MDL Number | MFCD00129791 |
| InChI Key | UUIVKBHZENILKB-UHFFFAOYSA-N |
| Synonym | dibromocyanoacetamide, dbnpa, 2,2-dibromo-3-nitrilopropionamide, acetamide, 2,2-dibromo-2-cyano, xd-7287l antimicrobial, 2-cyano-2,2-dibromoacetamide, 2,2-dibromo-2-carbamoylacetonitrile, caswell no. 287aa, unii-7n51qgl6mj, epa pesticide chemical code 101801 |
| PubChem CID | 25059 |
| IUPAC Name | 2,2-dibromo-2-cyanoacetamide |
| SMILES | NC(=O)C(Br)(Br)C#N |
Chemical Identifiers
| 10222-01-2 | |
| 241.87 | |
| UUIVKBHZENILKB-UHFFFAOYSA-N | |
| 25059 | |
| NC(=O)C(Br)(Br)C#N |
| C3H2Br2N2O | |
| MFCD00129791 | |
| dibromocyanoacetamide, dbnpa, 2,2-dibromo-3-nitrilopropionamide, acetamide, 2,2-dibromo-2-cyano, xd-7287l antimicrobial, 2-cyano-2,2-dibromoacetamide, 2,2-dibromo-2-carbamoylacetonitrile, caswell no. 287aa, unii-7n51qgl6mj, epa pesticide chemical code 101801 | |
| 2,2-dibromo-2-cyanoacetamide |
Specifications
| 10222-01-2 | |
| C3H2Br2N2O | |
| 25 g | |
| dibromocyanoacetamide, dbnpa, 2,2-dibromo-3-nitrilopropionamide, acetamide, 2,2-dibromo-2-cyano, xd-7287l antimicrobial, 2-cyano-2,2-dibromoacetamide, 2,2-dibromo-2-carbamoylacetonitrile, caswell no. 287aa, unii-7n51qgl6mj, epa pesticide chemical code 101801 | |
| NC(=O)C(Br)(Br)C#N | |
| 241.87 | |
| 241.87 | |
| Crystalline Powder |
| 125°C | |
| MFCD00129791 | |
| 3439 | |
| UUIVKBHZENILKB-UHFFFAOYSA-N | |
| 2,2-dibromo-2-cyanoacetamide | |
| 25059 | |
| ≥98.0% (N) | |
| 2,2-Dibromo-2-cyanoacetamide |
Safety and Handling
EINECSNumber : (2)-2795
RTECSNumber : AB5956000
TSCA : Yes