missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dibromo-2-cyanoacetamide 98.0+%, TCI America™

Catalog No. D290225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D290225G 25 g
D2902500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D290225G Supplier TCI America Supplier No. D290225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10222-01-2
Molecular Formula C3H2Br2N2O
Molecular Weight (g/mol) 241.87
MDL Number MFCD00129791
InChI Key UUIVKBHZENILKB-UHFFFAOYSA-N
Synonym dibromocyanoacetamide, dbnpa, 2,2-dibromo-3-nitrilopropionamide, acetamide, 2,2-dibromo-2-cyano, xd-7287l antimicrobial, 2-cyano-2,2-dibromoacetamide, 2,2-dibromo-2-carbamoylacetonitrile, caswell no. 287aa, unii-7n51qgl6mj, epa pesticide chemical code 101801
PubChem CID 25059
IUPAC Name 2,2-dibromo-2-cyanoacetamide
SMILES NC(=O)C(Br)(Br)C#N

Specifications

Melting Point 125°C
Quantity 25 g
UN Number 3439
Formula Weight 241.87
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Dibromo-2-cyanoacetamide
EINECSNumber (2)-2795
RTECSNumber AB5956000
TSCA Yes