missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dibromo-1,3-diphenyl-1,3-propanedione 98.0+%, TCI America™

Catalog No. D232410G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D232410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D232410G Supplier TCI America Supplier No. D232410G
Only null left

Chemical Identifiers

CAS 16619-55-9
Molecular Formula C15H10Br2O2
Molecular Weight (g/mol) 382.05
MDL Number MFCD00068642
InChI Key GSWSUDFFJVJMLG-UHFFFAOYSA-N
Synonym Dibenzoyldibromomethane
PubChem CID 12356677
IUPAC Name 2,2-dibromo-1,3-diphenylpropane-1,3-dione
SMILES BrC(Br)(C(=O)C1=CC=CC=C1)C(=O)C1=CC=CC=C1

Specifications

Color White
Quantity 10 g
Formula Weight 382.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Dibromo-1,3-diphenyl-1,3-propanedione