missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dibromo-1,1'-binaphthyl 96.0+%, TCI America™

Catalog No. D42211G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42211G 1 g
D42215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42211G Supplier TCI America Supplier No. D42211G

Chemical Identifiers

CAS 74866-28-7
Molecular Formula C20H12Br2
Molecular Weight (g/mol) 412.124
MDL Number MFCD00188005
InChI Key IJUDEFZBMMRSNM-UHFFFAOYSA-N
Synonym 2,2'-dibromo-1,1'-binaphthyl, 2,2'-dibromo-1,1'-binaphthalene, 1,1'-binaphthalene, 2,2'-dibromo, racemic-2,2'-dibromo-1,1'-binaphthyl, 1r-2,2'-dibromo-1,1'-binaphthalene, 1s-2,2'-dibromo-1,1'-binaphthalene, 2-bromo-1-2-bromonaphthalen-1-yl naphthalene, s---2,2-dibromo-1,1'-binaphthyl, 2,2'-dibromo 1,1' binaphthyl, 1,1'-bi 2-bromonaphthalene
PubChem CID 619909
IUPAC Name 2-bromo-1-(2-bromonaphthalen-1-yl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)Br)Br

Specifications

Melting Point 184°C
Quantity 1 g
Formula Weight 412.12
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dibromo-1,1′-binaphthyl