Learn More
2,2-Di-n-propylacetyl chloride, 98%
CAS: 2936-08-5 | C8H15ClO | 162.66 g/mol
$133.45 - $133.45
Chemical Identifiers
| CAS | 2936-08-5 |
|---|---|
| Molecular Formula | C8H15ClO |
| Molecular Weight (g/mol) | 162.66 |
| MDL Number | MFCD00051446 |
| InChI Key | PITHYUDHKJKJNQ-UHFFFAOYSA-N |
| Synonym | 2-propylvaleryl chloride, valproyl chloride, 2,2-di-n-propylacetyl chloride, dipropylacetyl chloride, 2-propylvaleroyl chloride, 2-n-propyl-n-valeroyl chloride, valproic acid chloride, di-n-propylacetyl chloride, acmc-2097cd, 2-n-propylvaleroyl chloride |
| PubChem CID | 76240 |
| IUPAC Name | 2-propylpentanoyl chloride |
| SMILES | CCCC(CCC)C(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0885206
|
Thermo Scientific Chemicals
L0885206 |
5 g |
Each for $133.45
|
|
|||||
Description
2,2-Di-n-propylacetyl chloride is used to produce 1,2-bis-(2-propyl-valerylamino)-ethan.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2-Di-n-propylacetyl chloride is used to produce 1,2-bis-(2-propyl-valerylamino)-ethan.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Incompatible materials are Oxidizing agents and bases. Store away from moisture and strong bases.
Chemical Identifiers
| 2936-08-5 | |
| 162.66 | |
| PITHYUDHKJKJNQ-UHFFFAOYSA-N | |
| 76240 | |
| CCCC(CCC)C(=O)Cl |
| C8H15ClO | |
| MFCD00051446 | |
| 2-propylvaleryl chloride, valproyl chloride, 2,2-di-n-propylacetyl chloride, dipropylacetyl chloride, 2-propylvaleroyl chloride, 2-n-propyl-n-valeroyl chloride, valproic acid chloride, di-n-propylacetyl chloride, acmc-2097cd, 2-n-propylvaleroyl chloride | |
| 2-propylpentanoyl chloride |
Specifications
| 2936-08-5 | |
| 68°C to 70°C (10 mmHg) | |
| MFCD00051446 | |
| UN3265 | |
| Moisture sensitive | |
| Reacts with water. | |
| CCCC(CCC)C(=O)Cl | |
| 162.66 | |
| 162.66 | |
| 2,2-Di-n-propylacetyl chloride |
| 0.95 | |
| C8H15ClO | |
| 5 g | |
| 1098856 | |
| 2-propylvaleryl chloride, valproyl chloride, 2,2-di-n-propylacetyl chloride, dipropylacetyl chloride, 2-propylvaleroyl chloride, 2-n-propyl-n-valeroyl chloride, valproic acid chloride, di-n-propylacetyl chloride, acmc-2097cd, 2-n-propylvaleroyl chloride | |
| PITHYUDHKJKJNQ-UHFFFAOYSA-N | |
| 2-propylpentanoyl chloride | |
| 76240 | |
| 98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 220-914-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only