missing translation for 'onlineSavingsMsg'
Learn More

2,2-Di-n-propylacetyl chloride, 98%

Catalog No. AAL0885206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0885206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0885206 Supplier Thermo Scientific Chemicals Supplier No. L0885206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2936-08-5 | C8H15ClO | 162.66 g/mol

2,2-Di-n-propylacetyl chloride is used to produce 1,2-bis-(2-propyl-valerylamino)-ethan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Di-n-propylacetyl chloride is used to produce 1,2-bis-(2-propyl-valerylamino)-ethan.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Incompatible materials are Oxidizing agents and bases. Store away from moisture and strong bases.

Chemical Identifiers

CAS 2936-08-5
Molecular Formula C8H15ClO
Molecular Weight (g/mol) 162.66
MDL Number MFCD00051446
InChI Key PITHYUDHKJKJNQ-UHFFFAOYSA-N
Synonym 2-propylvaleryl chloride, valproyl chloride, 2,2-di-n-propylacetyl chloride, dipropylacetyl chloride, 2-propylvaleroyl chloride, 2-n-propyl-n-valeroyl chloride, valproic acid chloride, di-n-propylacetyl chloride, acmc-2097cd, 2-n-propylvaleroyl chloride
PubChem CID 76240
IUPAC Name 2-propylpentanoyl chloride
SMILES CCCC(CCC)C(=O)Cl

Specifications

Density 0.95
Boiling Point 68°C to 70°C (10 mmHg)
Quantity 5 g
UN Number UN3265
Beilstein 1098856
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 162.66
Percent Purity 98%
Chemical Name or Material 2,2-Di-n-propylacetyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 220-914-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only