missing translation for 'onlineSavingsMsg'
Learn More

2,2-Di-n-octyl-1,3-propanediol 95.0+%, TCI America™

Catalog No. D28425G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28425G 5 g
D28421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28425G Supplier TCI America Supplier No. D28425G
Only null left

Chemical Identifiers

CAS 106868-09-1
Molecular Formula C19H40O2
Molecular Weight (g/mol) 300.53
MDL Number MFCD03844790
InChI Key NFPNQEAEXIXGNY-UHFFFAOYSA-N
Synonym 9,9-Bis(hydroxymethyl)heptadecane, 1,3-Dihydroxy-2,2-di-n-octylpropane
PubChem CID 10891950
IUPAC Name 2,2-dioctylpropane-1,3-diol
SMILES CCCCCCCCC(CO)(CO)CCCCCCCC

Specifications

Color White
Quantity 5 g
Formula Weight 300.53
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Di-n-octyl-1,3-propanediol