missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Di(2-tetrahydrofuryl)propane 96.0+%, TCI America™
$78.29 - $646.83
Chemical Identifiers
| CAS | 89686-69-1 |
|---|---|
| Molecular Formula | C11H20O2 |
| Molecular Weight (g/mol) | 184.279 |
| MDL Number | MFCD00142763 |
| InChI Key | FZLHAQMQWDDWFI-UHFFFAOYSA-N |
| PubChem CID | 551484 |
| IUPAC Name | 2-[2-(oxolan-2-yl)propan-2-yl]oxolane |
| SMILES | CC(C)(C1CCCO1)C2CCCO2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D203425ML
|
TCI America
D203425ML |
25 mL |
Each for $78.29
|
|
|||||
|
D2034100ML
|
TCI America
D2034100ML |
100 mL |
Each for $214.37
|
|
|||||
|
D2034500ML
|
TCI America
D2034500ML |
500 mL |
Each for $646.83
|
|
|||||
Chemical Identifiers
| 89686-69-1 | |
| 184.279 | |
| FZLHAQMQWDDWFI-UHFFFAOYSA-N | |
| 2-[2-(oxolan-2-yl)propan-2-yl]oxolane |
| C11H20O2 | |
| MFCD00142763 | |
| 551484 | |
| CC(C)(C1CCCO1)C2CCCO2 |
Specifications
| 89686-69-1 | |
| 146°C | |
| Clear Liquid at 20°C | |
| MFCD00142763 | |
| CC(C)(C1CCCO1)C2CCCO2 | |
| 184.279 | |
| 184.28 | |
| 2,2-Di(2-tetrahydrofuryl)propane |
| Colorless | |
| 25 mL | |
| C11H20O2 | |
| FZLHAQMQWDDWFI-UHFFFAOYSA-N | |
| 2-[2-(oxolan-2-yl)propan-2-yl]oxolane | |
| 551484 | |
| ≥96.0% (GC) |
Safety and Handling
EINECSNumber : (5)-6167
TSCA : Yes