missing translation for 'onlineSavingsMsg'
Learn More

2,2-Di(2-tetrahydrofuryl)propane 96.0+%, TCI America™

Catalog No. D203425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D203425ML 25 mL
D2034100ML 100 mL
D2034500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D203425ML Supplier TCI America Supplier No. D203425ML

Chemical Identifiers

CAS 89686-69-1
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00142763
InChI Key FZLHAQMQWDDWFI-UHFFFAOYSA-N
PubChem CID 551484
IUPAC Name 2-[2-(oxolan-2-yl)propan-2-yl]oxolane
SMILES CC(C)(C1CCCO1)C2CCCO2

Specifications

Color Colorless
Quantity 25 mL
Physical Form Clear Liquid at 20°C
Boiling Point 146°C
Formula Weight 184.28
Percent Purity ≥96.0% (GC)
Chemical Name or Material 2,2-Di(2-tetrahydrofuryl)propane
EINECSNumber (5)-6167
TSCA Yes