missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Cyanophenoxy)acetamide, 98%

Catalog No. p-7057002
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6627103 1 g
AAH6627106 5 g
AAH6627114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6627103 Supplier Thermo Scientific Chemicals Supplier No. H6627103
Only null left

CAS: 54802-12-9 | C9H8N2O2 | 176.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54802-12-9
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD00297091
InChI Key RYOYNMJXEHNYAG-UHFFFAOYSA-N
Synonym 2-2-cyanophenoxy acetamide, acetamide, 2-2-cyanophenoxy, 2-cyanophenoxyacetamide
PubChem CID 12353988
IUPAC Name 2-(2-cyanophenoxy)acetamide
SMILES C1=CC=C(C(=C1)C#N)OCC(=O)N

Specifications

Quantity 1 g
UN Number UN3439
Beilstein 2939357
Formula Weight 176.17
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-(2-Cyanophenoxy)acetamide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only