missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Chlorophenyl)glycine 98.0+%, TCI America™
$390.80 - $390.80
Chemical Identifiers
| CAS | 88744-36-9 |
|---|---|
| Molecular Formula | C8H8ClNO2 |
| Molecular Weight (g/mol) | 185.607 |
| MDL Number | MFCD00049324 |
| InChI Key | LMIZLNPFTRQPSF-UHFFFAOYSA-N |
| Synonym | 2-amino-2-2-chlorophenyl acetic acid, 2-2-chlorophenyl glycine, 2-chlorophenylglycine, dl-2-2-chlorophenyl glycine, amino 2-chlorophenyl acetic acid, amino-2-chloro-phenyl-acetic acid, dl-2-chlorophenylglycine, 2-chlorophenyl glycine, 2-chloro-dl-phenylglycine, h-dl-phg 2-cl-oh |
| PubChem CID | 290729 |
| IUPAC Name | 2-amino-2-(2-chlorophenyl)acetic acid |
| SMILES | C1=CC=C(C(=C1)C(C(=O)O)N)Cl |
Chemical Identifiers
| 88744-36-9 | |
| 185.607 | |
| LMIZLNPFTRQPSF-UHFFFAOYSA-N | |
| 290729 | |
| C1=CC=C(C(=C1)C(C(=O)O)N)Cl |
| C8H8ClNO2 | |
| MFCD00049324 | |
| 2-amino-2-2-chlorophenyl acetic acid, 2-2-chlorophenyl glycine, 2-chlorophenylglycine, dl-2-2-chlorophenyl glycine, amino 2-chlorophenyl acetic acid, amino-2-chloro-phenyl-acetic acid, dl-2-chlorophenylglycine, 2-chlorophenyl glycine, 2-chloro-dl-phenylglycine, h-dl-phg 2-cl-oh | |
| 2-amino-2-(2-chlorophenyl)acetic acid |
Specifications
| 88744-36-9 | |
| White | |
| MFCD00049324 | |
| 2-amino-2-2-chlorophenyl acetic acid, 2-2-chlorophenyl glycine, 2-chlorophenylglycine, dl-2-2-chlorophenyl glycine, amino 2-chlorophenyl acetic acid, amino-2-chloro-phenyl-acetic acid, dl-2-chlorophenylglycine, 2-chlorophenyl glycine, 2-chloro-dl-phenylglycine, h-dl-phg 2-cl-oh | |
| C1=CC=C(C(=C1)C(C(=O)O)N)Cl | |
| 185.607 | |
| 185.61 | |
| Crystalline Powder |
| 186°C | |
| C8H8ClNO2 | |
| 5 g | |
| LMIZLNPFTRQPSF-UHFFFAOYSA-N | |
| 2-amino-2-(2-chlorophenyl)acetic acid | |
| 290729 | |
| ≥98.0% (HPLC,T) | |
| 2-(2-Chlorophenyl)glycine |
Safety and Handling
TSCA : No