missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chlorophenyl)ethylamine 98.0+%, TCI America™

Catalog No. C138625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C138625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C138625G Supplier TCI America Supplier No. C138625G

Chemical Identifiers

CAS 13078-80-3
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00008185
InChI Key RZBOMSOHMOVUES-UHFFFAOYSA-N
Synonym 2-2-chlorophenyl ethylamine, 2-2-chlorophenyl ethanamine, 2-chlorophenethylamine, o-chlorophenethylamine, benzeneethanamine, 2-chloro, 2'-chlorophenethylamine, 1-amino-2-2-chlorophenyl ethane, 2-2-chloro-phenyl-ethylamine, 2-2-chlorophenyl ethan-1-amine, 2-chloro-2-phenethylamine
PubChem CID 83117
IUPAC Name 2-(2-chlorophenyl)ethanamine
SMILES C1=CC=C(C(=C1)CCN)Cl

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 g
Formula Weight 155.63
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Chlorophenyl)ethylamine
EINECSNumber (3)-3855
TSCA No