missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chlorophenyl)ethylamine, 97%

Catalog No. AC222840100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222840100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC222840100 Supplier Thermo Scientific Chemicals Supplier No. 222840100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13078-80-3 | C8H10ClN | 155.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13078-80-3
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD00008185
InChI Key RZBOMSOHMOVUES-UHFFFAOYSA-N
Synonym 2-2-chlorophenyl ethylamine, 2-2-chlorophenyl ethanamine, 2-chlorophenethylamine, o-chlorophenethylamine, benzeneethanamine, 2-chloro, 2'-chlorophenethylamine, 1-amino-2-2-chlorophenyl ethane, 2-2-chloro-phenyl-ethylamine, 2-2-chlorophenyl ethan-1-amine, 2-chloro-2-phenethylamine
PubChem CID 83117
IUPAC Name 2-(2-chlorophenyl)ethanamine
SMILES C1=CC=C(C(=C1)CCN)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1060g/mL
Boiling Point 120.0°C (15.0 mmHg)
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5505 to 1.5525
Linear Formula ClC6H4CH2CH2NH2
Quantity 10 g
Beilstein 12, III, 2431
Specific Gravity 1.106
Formula Weight 155.63
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(2-Chlorophenyl)ethylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 235-982-1

RUO – Research Use Only