missing translation for 'onlineSavingsMsg'
Learn More

2'-(2-Chloroanilino)-6'-(dibutylamino)fluoran 97.0+%, TCI America™

Catalog No. p-7125207
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C194625G 25 g
C1946500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C194625G Supplier TCI America Supplier No. C194625G
Only null left

Chemical Identifiers

CAS 82137-81-3
Molecular Formula C34H33ClN2O3
Molecular Weight (g/mol) 553.10
MDL Number MFCD08276326
InChI Key ZXYPDMNPLWGWBI-UHFFFAOYNA-N
PubChem CID 13668872
IUPAC Name 2'-[(2-chlorophenyl)amino]-6'-(dibutylamino)-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
SMILES CCCCN(CCCC)C1=CC2=C(C=C1)C1(OC(=O)C3=CC=CC=C13)C1=C(O2)C=CC(NC2=CC=CC=C2Cl)=C1

Specifications

Melting Point 186°C
Color Red-White
Quantity 25 g
Synonym 2′C-(2-Chlorophenylamino)-6′C-(dibutylamino)fluoran
Formula Weight 553.10
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2′-(2-Chloroanilino)-6′-(dibutylamino)fluoran
EINECSNumber (4)-1408&(5)-5678
TSCA No