missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Butoxyethoxy)ethanol, 99+%

Catalog No. p-2689606
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149710010 1 L Glass bottle
AC149710025 2.5 L Glass bottle
AC149710100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC149710010 Supplier Thermo Scientific Chemicals Supplier No. 149710010
Only null left

CAS: 112-34-5 | C8H18O3 | 162.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-34-5
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.23
InChI Key OAYXUHPQHDHDDZ-UHFFFAOYSA-N
Synonym 2-2-butoxyethoxy ethanol, diethylene glycol monobutyl ether, butyldiglycol, butyl carbitol, diethylene glycol butyl ether, butyl diglycol, butyl digol, ethanol, 2-2-butoxyethoxy, butoxyethoxyethanol, butoxydiglycol
PubChem CID 8177
IUPAC Name 2-(2-butoxyethoxy)ethanol
SMILES CCCCOCCOCCO

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -68.0°C
Density 0.9550g/mL
Boiling Point 231.0°C
Flash Point 100°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4310 to 1.4330
Linear Formula CH3(CH2)3OCH2CH2OCH2CH2OH
Quantity 1 L
Beilstein 01,IV,2394
Merck Index 15,3133
Specific Gravity 0.955
Solubility Information Solubility in water: soluble. Other solubilities: miscible with oils, miscible with most common organic solvents
Viscosity 6.16 mPa.s (20°C)
Formula Weight 162.23
Percent Purity 99+%
Chemical Name or Material 2-(2-Butoxyethoxy)ethanol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-961-6

RUO – Research Use Only