missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Butoxyethoxy)ethanol, 99%, Spectrum™ Chemical

Catalog No. p-7165161
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608134 1 L Amber Glass Bottle
18608137 4 L Amber Glass Bottle
18608135 20 L Steel Can
18608136 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608134 Supplier Spectrum Chemical Mfg Cor Supplier No. B11781LTGL
Only null left

Chemical Identifiers

CAS 112-34-5
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.23
InChI Key OAYXUHPQHDHDDZ-UHFFFAOYSA-N
IUPAC Name 2-(2-butoxyethoxy)ethan-1-ol
SMILES CCCCOCCOCCO

Specifications

Acidity 0.0001
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 L
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.