missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromophenyl)pyridine 98.0+%, TCI America™

Catalog No. B5044200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5044200MG 200 mg
B50441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5044200MG Supplier TCI America Supplier No. B5044200MG

Chemical Identifiers

CAS 109306-86-7
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.096
MDL Number MFCD15529755
InChI Key NNGXNALKPNFUQS-UHFFFAOYSA-N
PubChem CID 13223929
IUPAC Name 2-(2-bromophenyl)pyridine
SMILES C1=CC=C(C(=C1)C2=CC=CC=N2)Br

Specifications

Color Yellow
Boiling Point 139°C
Quantity 200 mg
Formula Weight 234.10
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2-Bromophenyl)pyridine