missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromophenyl)ethylamine 98.0+%, TCI America™

Catalog No. p-7123258
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B39385G 5 g
B393825G 25 g
2 options

Catalog No. B39385G

Supplier: TCI America B39385G

Only null left

Chemical Identifiers

CAS 65185-58-2
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD01529874
InChI Key ITRNQMJXZUWZQL-UHFFFAOYSA-N
Synonym 2-bromophenethylamine, 2-2-bromophenyl ethanamine, 2-2-bromophenyl ethylamine, 2-2-bromophenyl ethan-1-amine, benzeneethanamine, 2-bromo, 2-bromo phenethylamin, 2-2-bromopheneyl ethylamine, 1-amino-2-bromophenyl ethane, 2-bromo phenethylamine, 2-bromo-phenethylamine
PubChem CID 2734091
IUPAC Name 2-(2-bromophenyl)ethanamine
SMILES C1=CC=C(C(=C1)CCN)Br

Specifications

Color Yellow
Boiling Point 85°C
Quantity 5 g
UN Number 2735
Formula Weight 200.08
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-(2-Bromophenyl)ethylamine