missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromophenyl)ethylamine, 97%

Catalog No. AAH6073909
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6073909 10 g
AAH6073904 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6073909 Supplier Thermo Scientific Chemicals Supplier No. H6073909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 65185-58-2 | C8H10BrN | 200.079 g/mol

2-(2-Bromophenyl)ethylamine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Bromophenyl)ethylamine is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 65185-58-2
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD01529874
InChI Key ITRNQMJXZUWZQL-UHFFFAOYSA-N
Synonym 2-bromophenethylamine, 2-2-bromophenyl ethanamine, 2-2-bromophenyl ethylamine, 2-2-bromophenyl ethan-1-amine, benzeneethanamine, 2-bromo, 2-bromo phenethylamin, 2-2-bromopheneyl ethylamine, 1-amino-2-bromophenyl ethane, 2-bromo phenethylamine, 2-bromo-phenethylamine
PubChem CID 2734091
IUPAC Name 2-(2-bromophenyl)ethanamine
SMILES C1=CC=C(C(=C1)CCN)Br

Specifications

Density 1.396
Boiling Point 253°C
Refractive Index 1.577
Quantity 10 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 200.08
Percent Purity 97%
Chemical Name or Material 2-(2-Bromophenyl)ethylamine
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only