missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Bromomethyl)phenyl]thiophene, ≥95%, Thermo Scientific™

Catalog No. CC35508CB
Change view
Click to view available options
Amber Glass Bottle; 250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC35508CB Amber Glass Bottle; 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC35508CB Supplier Thermo Scientific Chemicals Supplier No. CC35508CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 791078-04-1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Molecular Formula C11H9BrS
Quantity 250mg
MDL Number MFCD07368537
Synonym 2-2-bromomethyl phenyl thiophene, 2-thien-2-yl benzyl bromide, thiophene,2-2-bromomethyl phenyl, 1-bromomethyl-2-thien-2-yl benzene, thiophene, 2-2-bromomethyl phenyl
InChI Key IJNVNLWIQXMBPA-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)CBr)C2=CC=CS2
IUPAC Name 2-[2-(bromomethyl)phenyl]thiophene
Molecular Weight (g/mol) 253.157
PubChem CID 7060555
Formula Weight 253.16
Percent Purity 95%
Chemical Name or Material 2-[2-(Bromomethyl)phenyl]thiophene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.