Learn More
2-(2-Bromoethyl)-1,3-dioxane, 98%
CAS: 33884-43-4 | C6H11BrO2 | 195.06 g/mol
Supplier: Thermo Scientific Chemicals L0095818
Description
2-(2-Bromoethyl)-1,3-dioxane, Provides a useful means of introducing a masked β-formylethyl group. In comparison with the corresponding dioxolane (following entry), the dioxane is more stable to acid, and the Grignard is thermally stable. Reaction of the Grignard with an acid chloride gives the ketone in high yield without further reaction to the tertiary alcohol. Subsequent hydrolysis of the 1,3-dioxane gives the -ketoaldehyde. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(2-Bromoethyl)-1,3-dioxane, Provides a useful means of introducing a masked Ã-formylethyl group. In comparison with the corresponding dioxolane (following entry), the dioxane is more stable to acid, and the Grignard is thermally stable. Reaction of the Grignard with an acid chloride gives the ketone in high yield without further reaction to the tertiary alcohol. Subsequent hydrolysis of the 1,3-dioxane gives the -ketoaldehyde. It is also used as a pharmaceutical intermediate.
Solubility
It is not soluble in water but soluble in organic solvent such as ethanol.
Notes
Air, Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
| 2-(2-Bromoethyl)-1,3-dioxane | |
| 1.434 | |
| 96°C (204°F) | |
| 1.482 | |
| 50 g | |
| 2-2-bromoethyl-1,3-dioxane, 1,3-dioxane, 2-2-bromoethyl, 3-bromopropionaldehyde trimethylene acetal, 2 2-bromoethyl-1,3-dioxane, 2-2-bromoethyl-1,3 dioxane, bromoethyl-1,3-dioxane, acmc-209i3o, 2-2-bromoetyl 1,3-dioxane, wmdhqehpovoeog-uhfffaoysa, 2-2-bromoethyl 1,3-dioxane | |
| KXMZOKKPQZRPRN-UHFFFAOYNA-N | |
| 2-(1-bromoethyl)-1,3-dioxane | |
| 520656 | |
| 98% |
| 33884-43-4 | |
| 67°C to 70°C (3 mmHg) | |
| C6H11BrO2 | |
| MFCD00006567 | |
| 1421628 | |
| It is not soluble in water but soluble in organic solvent such as ethanol. | |
| CC(Br)C1OCCCO1 | |
| 195.06 | |
| 195.06 |
Chemical Identifiers
| 33884-43-4 | |
| 195.06 | |
| KXMZOKKPQZRPRN-UHFFFAOYNA-N | |
| 520656 |
| C6H11BrO2 | |
| MFCD00006567 | |
| 2-2-bromoethyl-1,3-dioxane, 1,3-dioxane, 2-2-bromoethyl, 3-bromopropionaldehyde trimethylene acetal, 2 2-bromoethyl-1,3-dioxane, 2-2-bromoethyl-1,3 dioxane, bromoethyl-1,3-dioxane, acmc-209i3o, 2-2-bromoetyl 1,3-dioxane, wmdhqehpovoeog-uhfffaoysa, 2-2-bromoethyl 1,3-dioxane | |
| CC(Br)C1OCCCO1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H341
EINECSNumber : 251-716-7
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only