missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Bromoethoxy)tetrahydro-2H-pyran (stabilized with K2CO3) 95.0+%, TCI America™
$199.46 - $704.62
Chemical Identifiers
| CAS | 17739-45-6 |
|---|---|
| Molecular Formula | C7H13BrO2 |
| Molecular Weight (g/mol) | 209.083 |
| InChI Key | GCUOLJOTJRUDIZ-UHFFFAOYSA-N |
| Synonym | 2-2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy oxane, 2h-pyran, 2-2-bromoethoxy tetrahydro, 2-2-bromo-ethoxy-tetrahydro-pyran, 2-2-bromoethoxy tetrahydropyran, 2 2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy-tetrahydro-2h-pyran, 2h-pyran,3-2-bromoethoxy tetrahydro, 2-2-bromoethyl oxy tetrahaydro-2h-pyran |
| PubChem CID | 86621 |
| IUPAC Name | 2-(2-bromoethoxy)oxane |
| SMILES | C1CCOC(C1)OCCBr |
Chemical Identifiers
| 17739-45-6 | |
| 209.083 | |
| 2-2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy oxane, 2h-pyran, 2-2-bromoethoxy tetrahydro, 2-2-bromo-ethoxy-tetrahydro-pyran, 2-2-bromoethoxy tetrahydropyran, 2 2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy-tetrahydro-2h-pyran, 2h-pyran,3-2-bromoethoxy tetrahydro, 2-2-bromoethyl oxy tetrahaydro-2h-pyran | |
| 2-(2-bromoethoxy)oxane |
| C7H13BrO2 | |
| GCUOLJOTJRUDIZ-UHFFFAOYSA-N | |
| 86621 | |
| C1CCOC(C1)OCCBr |
Specifications
| 17739-45-6 | |
| 75°C | |
| 5 g | |
| GCUOLJOTJRUDIZ-UHFFFAOYSA-N | |
| 2-(2-bromoethoxy)oxane | |
| 86621 | |
| ≥95.0% (GC) | |
| 2-(2-Bromoethoxy)tetrahydro-2H-pyran (stabilized with K2CO3) |
| Yellow | |
| C7H13BrO2 | |
| 2-2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy oxane, 2h-pyran, 2-2-bromoethoxy tetrahydro, 2-2-bromo-ethoxy-tetrahydro-pyran, 2-2-bromoethoxy tetrahydropyran, 2 2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy-tetrahydro-2h-pyran, 2h-pyran,3-2-bromoethoxy tetrahydro, 2-2-bromoethyl oxy tetrahaydro-2h-pyran | |
| C1CCOC(C1)OCCBr | |
| 209.083 | |
| 209.08 | |
| Liquid |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator