missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)-1,1,1,2,3,3-hexafluoro-3-(pentafluoroethoxy)propane 98.0+%, TCI America™

Catalog No. B41681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41681G 1 g
B41685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41681G Supplier TCI America Supplier No. B41681G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1482416-44-3
Molecular Formula C7BrF15O2
Molecular Weight (g/mol) 480.96
MDL Number MFCD27918479
InChI Key CGHGTHHVJHGUNI-UHFFFAOYNA-N
PubChem CID 75529904
IUPAC Name 2-(2-bromo-1,1,2,2-tetrafluoroethoxy)-1,1,1,2,3,3-hexafluoro-3-(1,1,2,2,2-pentafluoroethoxy)propane
SMILES FC(F)(F)C(F)(F)OC(F)(F)C(F)(OC(F)(F)C(F)(F)Br)C(F)(F)F

Specifications

Color Colorless
Quantity 1 g
Formula Weight 480.96
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)-1,1,1,2,3,3-hexafluoro-3-(pentafluoroethoxy)propane