Learn More
2,2'-Bithiophene, 98%
CAS: 492-97-7 | C8H6S2 | 166.256 g/mol
Supplier: Thermo Scientific Chemicals A1233506
Description
2,2'-Bithiophene is used as a precursor in the preparation of 5,5'-bis(trimethylstannyl)-2,2'-bithiophene , which is used for the synthesis of small molecules and polymer semiconductors for organic field-effect transistor (OFETs), organic light-emitting diode (OLED), Plastic Light Emitting Diode (PLED) and Organic Photovoltaic (OPV) applications. It is associated with aryl iodides and involved in rhodium catalyzed arylation of heteroarenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2′-Bithiophene is used as a precursor in the preparation of 5,5′-bis(trimethylstannyl)-2,2′-bithiophene , which is used for the synthesis of small molecules and polymer semiconductors for organic field-effect transistor (OFETs), organic light-emitting diode (OLED), Plastic Light Emitting Diode (PLED) and Organic Photovoltaic (OPV) applications. It is associated with aryl iodides and involved in rhodium catalyzed arylation of heteroarenes.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Keep container tightly closed in a dry and well-ventilated place.
Specifications
| 2,2'-Bithiophene | |
| 30°C to 33°C | |
| >110°C (230°F) | |
| MFCD00005414 | |
| 3039 | |
| 2,2'-bithiophene, 2,2'-bithienyl, dithienyl, bithiophene, 2,2'-dithienyl, 2,2' bithiophenyl, 2,2-bithiophene, 2,2'-dithiophene, alpha-bithiophene, 2-2-thienyl thiophene | |
| OHZAHWOAMVVGEL-UHFFFAOYSA-N | |
| 2-thiophen-2-ylthiophene | |
| 68120 | |
| 166.27 |
| 492-97-7 | |
| 260°C | |
| C8H6S2 | |
| 5 g | |
| Light sensitive | |
| Insoluble in water. | |
| C1=CSC(=C1)C2=CC=CS2 | |
| 166.256 | |
| CHEBI:36821 | |
| 98% |
Chemical Identifiers
| 492-97-7 | |
| 166.256 | |
| OHZAHWOAMVVGEL-UHFFFAOYSA-N | |
| 68120 | |
| 2-thiophen-2-ylthiophene |
| C8H6S2 | |
| MFCD00005414 | |
| 2,2'-bithiophene, 2,2'-bithienyl, dithienyl, bithiophene, 2,2'-dithienyl, 2,2' bithiophenyl, 2,2-bithiophene, 2,2'-dithiophene, alpha-bithiophene, 2-2-thienyl thiophene | |
| CHEBI:36821 | |
| C1=CSC(=C1)C2=CC=CS2 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-767-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only