missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Bithiophene 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 260°C |
| Quantity | 1 g |
| Formula Weight | 166.26 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 2,2′-Bithiophene |
Safety and Handling
| TSCA | Yes |