missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bithiophene 98.0+%, TCI America™

Catalog No. B12761G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12761G 1 g
B127625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12761G Supplier TCI America Supplier No. B12761G

Chemical Identifiers

CAS 492-97-7
Molecular Formula C8H6S2
Molecular Weight (g/mol) 166.256
MDL Number MFCD00005414
InChI Key OHZAHWOAMVVGEL-UHFFFAOYSA-N
Synonym 2,2'-bithiophene, 2,2'-bithienyl, dithienyl, bithiophene, 2,2'-dithienyl, 2,2' bithiophenyl, 2,2-bithiophene, 2,2'-dithiophene, alpha-bithiophene, 2-2-thienyl thiophene
PubChem CID 68120
ChEBI CHEBI:36821
IUPAC Name 2-thiophen-2-ylthiophene
SMILES C1=CSC(=C1)C2=CC=CS2

Specifications

Boiling Point 260°C
Quantity 1 g
Formula Weight 166.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,2′-Bithiophene