missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bithiophene, 97%

Catalog No. AC402690025
Change view
Click to view available options
Quantity:
2.5 g
20 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402690025 2.5 g Glass bottle
AC402690200 20 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC402690025 Supplier Thermo Scientific Chemicals Supplier No. 402690025
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 492-97-7 | C8H6S2 | 166.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 492-97-7
Molecular Formula C8H6S2
Molecular Weight (g/mol) 166.27
MDL Number MFCD00005414
InChI Key OHZAHWOAMVVGEL-UHFFFAOYSA-N
Synonym 2,2'-bithiophene, 2,2'-bithienyl, dithienyl, bithiophene, 2,2'-dithienyl, 2,2' bithiophenyl, 2,2-bithiophene, 2,2'-dithiophene, alpha-bithiophene, 2-2-thienyl thiophene
PubChem CID 68120
ChEBI CHEBI:36821
IUPAC Name 2-thiophen-2-ylthiophene
SMILES C1=CSC(=C1)C2=CC=CS2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 30.0°C to 35.0°C
Boiling Point 260.0°C
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Beilstein 19, 32
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in methanol
Formula Weight 166.27
Percent Purity 97%
Chemical Name or Material 2, 2'-Bithiophene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 207-767-2
TSCA TSCA

RUO – Research Use Only