missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bithiophene-5-carboxaldehyde 98.0+%, TCI America™

Catalog No. B18381G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18381G 1 g
B18385G 5 g
B183825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B18381G Supplier TCI America Supplier No. B18381G

Chemical Identifiers

CAS 3779-27-9
Molecular Formula C9H6OS2
Molecular Weight (g/mol) 194.266
MDL Number MFCD00052294
InChI Key FYBWRAXKYXTOQC-UHFFFAOYSA-N
Synonym 2,2'-bithiophene-5-carboxaldehyde, 2,2'-bithiophene-5-carbaldehyde, 2,2'-bithienyl-5-carboxaldehyde, 5-formyl-2,2'-bithiophene, 5-thiophen-2-yl thiophene-2-carbaldehyde, 5-2-thienyl thiophene-2-carbaldehyde, 2,2-bithiophene-5-carboxaldehyde
PubChem CID 364428
IUPAC Name 5-thiophen-2-ylthiophene-2-carbaldehyde
SMILES C1=CSC(=C1)C2=CC=C(S2)C=O

Specifications

Melting Point 57°C
Boiling Point 210°C
Quantity 1 g
Formula Weight 194.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bithiophene-5-carboxaldehyde