missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bithiophene-5,5'-dicarboxaldehyde 98.0+%, TCI America™

Catalog No. B53585G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53585G 5 g
B53581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53585G Supplier TCI America Supplier No. B53585G

Chemical Identifiers

CAS 32364-72-0
Molecular Formula C10H6O2S2
Molecular Weight (g/mol) 222.276
MDL Number MFCD00839981
InChI Key RXAXZMANGDHIJX-UHFFFAOYSA-N
Synonym 5,5′-Diformyl-2,2′-bithiophene
PubChem CID 2063643
IUPAC Name 5-(5-formylthiophen-2-yl)thiophene-2-carbaldehyde
SMILES C1=C(SC(=C1)C2=CC=C(S2)C=O)C=O

Specifications

Melting Point 215°C
Quantity 5 g
Formula Weight 222.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bithiophene-5,5′-dicarboxaldehyde