missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(hydroxymethyl)propionic acid, 98+%

Catalog No. AAL1401422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1401422 100 g
AAL1401436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1401422 Supplier Thermo Scientific Chemicals Supplier No. L1401422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4767-03-7 | C5H10O4 | 134.13 g/mol

2,2-Bis(hydroxymethyl)propionic acid is used in the synthesis and characterization studies of polyurethane dispersion modified with polymethylsiloxane (Si-PUD) and its nanocomposites incorporated with clays1.2,2-Bis(hydroxymethyl)propionic acid based dendrimers have potent applications as delivery systems for potent anticancer drugs such as cisplatin and doxorubicin, as well as agents for both boron neutron capture therapy and photodynamic therapy Simple derivatives of this acid have been used to construct dendritic architectures via the click azide-acetylene cycloaddition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in acetone (slightly), methanol, and water. Insoluble in benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature.

Chemical Identifiers

CAS 4767-03-7
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
MDL Number MFCD00004199
InChI Key PTBDIHRZYDMNKB-UHFFFAOYSA-N
Synonym 2,2-bis hydroxymethyl propionic acid, dimethylolpropionic acid, 3-hydroxy-2-hydroxymethyl-2-methylpropanoic acid, 2,2-bis hydroxymethyl propanoic acid, propanoic acid, 3-hydroxy-2-hydroxymethyl-2-methyl, 2,2-dimethylolpropionic acid, dimethylol propionic acid, 2,2-bis hydroxymethyl propanoicacid, unii-4nhi8v17mn, 4nhi8v17mn
PubChem CID 78501
IUPAC Name 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
SMILES CC(CO)(CO)C(=O)O

Specifications

Melting Point 187°C to 191°C
Quantity 100 g
Beilstein 1756385
Sensitivity Hygroscopic
Merck Index 14,3253
Solubility Information Soluble in acetone (slightly),methanol,and water. Insoluble in benzene.
Formula Weight 134.13
Percent Purity ≥98%
Chemical Name or Material 2,2-Bis(hydroxymethyl)propionic acid
Hazard Category H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 225-306-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only