missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%, for biochemistry

CAS: 6976-37-0 | C8H19NO5 | 209.24 g/mol

$255.68 - $255.68

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.24
MDL Number MFCD00002853
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
Synonym bis-tris, bistris, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis-tris buffer, 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, unii-q1xc3631cp, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl iminotris hydroxymethyl methane
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC327721000
SDS View Documents
Thermo Scientific Chemicals
327721000
100 g
Each for $255.68
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

6976-37-0
209.24
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C8H19NO5
MFCD00002853
bis-tris, bistris, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis-tris buffer, 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, unii-q1xc3631cp, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl iminotris hydroxymethyl methane
CHEBI:41250
C(CO)N(CCO)C(CO)(CO)CO

Specifications

6976-37-0
(0.1M in H2O), (0.1 M in H2O) at 0nm, 0.02 max. at 340nm, 0.04 max. at 280nm
103.0°C to 104.0°C
8.8 to 11.0 (1% aq. soln. at 25°C)
99+%
C8H19NO5
MFCD00002853
bis-tris, bistris, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis-tris buffer, 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, unii-q1xc3631cp, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl iminotris hydroxymethyl methane
OWMVSZAMULFTJU-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
81462
209.24
Biochemistry
2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol
99
100
Yellow to Orange
Authentic
Plastic bottle
(HOCH2CH2)2NC(CH2OH)3
100 g
Solubility in water: soluble. Other solubilities: soluble in alcohol
C(CO)N(CCO)C(CO)(CO)CO
209.24
CHEBI:41250
99+%
Solution
Videos
Safety and Handling

Safety and Handling

Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation

Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)

EINECSNumber : 230-237-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only