missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(allyloxymethyl)-1-butanol (contains Mono- and Tri-substituted Product) 85.0+%, TCI America™

Catalog No. B3003500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3003500G 500 g
B300325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3003500G Supplier TCI America Supplier No. B3003500G
Only null left

Chemical Identifiers

CAS 682-09-7
Molecular Formula C12H22O3
Molecular Weight (g/mol) 214.305
MDL Number MFCD00035791
InChI Key BDKDHWOPFRTWPP-UHFFFAOYSA-N
Synonym Trimethylolpropane Diallyl Ether
PubChem CID 61196
IUPAC Name 2,2-bis(prop-2-enoxymethyl)butan-1-ol
SMILES CCC(CO)(COCC=C)COCC=C

Specifications

Color Yellow
Boiling Point 122°C
Quantity 500 g
Formula Weight 214.31
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Bis(allyloxymethyl)-1-butanol (contains Mono- and Tri-substituted Product)
EINECSNumber (2)-0427
RTECSNumber TY6667000
TSCA Yes