missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(allyloxymethyl)-1-butanol (contains Mono- and Tri-substituted Product) 85.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 122°C |
| Quantity | 25 g |
| Formula Weight | 214.31 |
| Percent Purity | ≥85.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,2-Bis(allyloxymethyl)-1-butanol (contains Mono- and Tri-substituted Product) |
Safety and Handling
| EINECSNumber | (2)-0427 |
| RTECSNumber | TY6667000 |
| TSCA | Yes |