missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(4-hydroxycyclohexyl)propane (mixture of isomers) 93.0+%, TCI America™
$48.46 - $126.76
Chemical Identifiers
| CAS | 80-04-6 |
|---|---|
| Molecular Formula | C15H28O2 |
| Molecular Weight (g/mol) | 240.387 |
| MDL Number | MFCD00019334 |
| InChI Key | CDBAMNGURPMUTG-UHFFFAOYSA-N |
| PubChem CID | 94932 |
| IUPAC Name | 4-[2-(4-hydroxycyclohexyl)propan-2-yl]cyclohexan-1-ol |
| SMILES | CC(C)(C1CCC(CC1)O)C2CCC(CC2)O |
Chemical Identifiers
| 80-04-6 | |
| 240.387 | |
| CDBAMNGURPMUTG-UHFFFAOYSA-N | |
| 4-[2-(4-hydroxycyclohexyl)propan-2-yl]cyclohexan-1-ol |
| C15H28O2 | |
| MFCD00019334 | |
| 94932 | |
| CC(C)(C1CCC(CC1)O)C2CCC(CC2)O |
Specifications
| 80-04-6 | |
| C15H28O2 | |
| 25 g | |
| CDBAMNGURPMUTG-UHFFFAOYSA-N | |
| 4-[2-(4-hydroxycyclohexyl)propan-2-yl]cyclohexan-1-ol | |
| 94932 | |
| ≥93.0% (GC) | |
| 2,2-Bis(4-hydroxycyclohexyl)propane (mixture of isomers) |
| White | |
| MFCD00019334 | |
| p,p′C-Dihydroxy-2,2-dicyclohexylpropane, Perhydrobisphenol A | |
| CC(C)(C1CCC(CC1)O)C2CCC(CC2)O | |
| 240.387 | |
| 240.39 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0201
TSCA : Yes