missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,2'-biimidazole 97.0+%, TCI America™

Catalog No. B122525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B122525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B122525G Supplier TCI America Supplier No. B122525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

[Photopolymerization Initiator]

Chemical Identifiers

CAS 7189-82-4
Molecular Formula C42H28Cl2N4
Molecular Weight (g/mol) 659.61
MDL Number MFCD09028023
InChI Key MHDULSOPQSUKBQ-UHFFFAOYSA-N
PubChem CID 81610
IUPAC Name 2,2'-bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-2'H-1,2'-biimidazole
SMILES ClC1=CC=CC=C1C1=NC(=C(N1C1(N=C(C(=N1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1Cl)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 199°C
Color Yellow
Quantity 25 g
Formula Weight 659.61
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bis(2-chlorophenyl)-4,4′,5,5′-tetraphenyl-1,2′-biimidazole [Photopolyme
EINECSNumber (5)-5781
TSCA Yes