missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Biquinoline 98.0+%, TCI America™

Catalog No. p-7118465
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04701G 1 g
B04705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B04701G Supplier TCI America Supplier No. B04701G
Only null left

Chemical Identifiers

CAS 119-91-5
Molecular Formula C18H12N2
Molecular Weight (g/mol) 256.308
MDL Number MFCD00006740
InChI Key WPTCSQBWLUUYDV-UHFFFAOYSA-N
Synonym 2,2'-biquinoline, cuproin, cuproine, 2,2'-diquinolyl, 2,2'-biquinolyl, 2,2'-quinolyl, 2-2-quinolyl quinoline, 2,2-biquinoline, 2,2-diquinolyl, 2,2;-biquinoline
PubChem CID 8412
IUPAC Name 2-quinolin-2-ylquinoline
SMILES C1=CC=C2C(=C1)C=CC(=N2)C3=NC4=CC=CC=C4C=C3

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 1 g
Formula Weight 256.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Biquinoline