missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bipyrimidyl 95.0+%, TCI America™

Catalog No. p-7119681
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2496200MG 200 mg
B24961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2496200MG Supplier TCI America Supplier No. B2496200MG

Chemical Identifiers

CAS 34671-83-5
Molecular Formula C8H6N4
Molecular Weight (g/mol) 158.16
MDL Number MFCD00014600
InChI Key HKOAFLAGUQUJQG-UHFFFAOYSA-N
Synonym 2,2'-bipyrimidine, 2,2'-bipyrimidyl, 2,2-bipyrimidine, bpym, 2-pyrimidin-2-yl pyrimidine, bipyrimidine, 2,2'-bipyrimidinyl, 2,2'-dipyrimidyl, 2,2;-bipyrimidine
PubChem CID 123444
IUPAC Name 2,2'-bipyrimidine
SMILES C1=CN=C(N=C1)C1=NC=CC=N1

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 200 mg
Formula Weight 158.16
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bipyrimidyl