missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bipyrimidine, 96%

Catalog No. p-7043538
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL01190MD 250 mg
AAL0119003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL01190MD Supplier Thermo Scientific Chemicals Supplier No. L01190MD
Only null left

CAS: 34671-83-5 | C8H6N4 | 158.16 g/mol

2,2'-Bipyrimidine is used in the preparation of 5-bromo-[2,2']bipyrimidinyl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Bipyrimidine is used in the preparation of 5-bromo-[2,2′]bipyrimidinyl.

Solubility
Soluble in methanol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 34671-83-5
Molecular Formula C8H6N4
Molecular Weight (g/mol) 158.16
MDL Number MFCD00014600
InChI Key HKOAFLAGUQUJQG-UHFFFAOYSA-N
Synonym 2,2'-bipyrimidine, 2,2'-bipyrimidyl, 2,2-bipyrimidine, bpym, 2-pyrimidin-2-yl pyrimidine, bipyrimidine, 2,2'-bipyrimidinyl, 2,2'-dipyrimidyl, 2,2;-bipyrimidine
PubChem CID 123444
SMILES C1=CN=C(N=C1)C1=NC=CC=N1

Specifications

Melting Point 110°C to 116°C
Quantity 250 mg
Beilstein 607396
Solubility Information Soluble in methanol.
IUPAC Name 2-pyrimidin-2-ylpyrimidine
Formula Weight 158.16
Percent Purity 96%
Chemical Name or Material 2,2'-Bipyrimidine
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 252-137-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only