missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bipyridyl 99.0+%, TCI America™

Catalog No. B046825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B046825G 25 g
B0468100G 100 g
B0468500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B046825G Supplier TCI America Supplier No. B046825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 366-18-7 | C10H8N2 | 156.19 g/mol

Chemical Identifiers

CAS 366-18-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006212
InChI Key ROFVEXUMMXZLPA-UHFFFAOYSA-N
Synonym 2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
PubChem CID 1474
ChEBI CHEBI:30351
IUPAC Name 2,2'-bipyridine
SMILES C1=CC=C(N=C1)C1=CC=CC=N1

Specifications

Melting Point 72°C
Color White-Yellow
Boiling Point 273°C
Quantity 25 g
UN Number 2811
Formula Weight 156.19
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bipyridyl
EINECSNumber (5)-3723
RTECSNumber DW1750000
TSCA Yes