missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bipyridyl 1,1'-Dioxide 98.0+%, TCI America™

Catalog No. B38595G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B38595G Supplier TCI America Supplier No. B38595G

Chemical Identifiers

CAS 7275-43-6
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00065169
InChI Key FERMVCULDZOVOJ-KTKRTIGZSA-N
Synonym 2,2'-bipyridine 1,1'-dioxide, 2,2'-dipyridyl n,n'-dioxide, 2,2'-bipyridine n,n'-dioxide, 2,2'-dipyridyl-n,n'-dioxide
PubChem CID 6005843
IUPAC Name (Z)-1-oxo-1H,1'H-1λ⁵-[2,2'-bipyridinyliden]-1-ylium-1'-olate
SMILES [O-]N1C=CC=C\C1=C1/C=CC=C[N+]1=O

Specifications

Melting Point 311°C
Color White
Quantity 5 g
Formula Weight 188.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bipyridyl 1,1′-Dioxide
RTECSNumber DW1770000
TSCA No