missing translation for 'onlineSavingsMsg'
Learn More

(2,2'-Bipyridine)bis(2-phenylpyridinato)iridium(III) Hexafluorophosphate 90.0+%, TCI America™

Catalog No. b4893200mg
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4893200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4893200MG Supplier TCI America Supplier No. B4893200MG

Chemical Identifiers

CAS 106294-60-4
Molecular Formula C32H24F6IrN4P
Molecular Weight (g/mol) 801.753
InChI Key RJJGJTKSOSSNNL-UHFFFAOYSA-N
PubChem CID 54589530
IUPAC Name iridium(3+);2-phenylpyridine;2-pyridin-2-ylpyridine;hexafluorophosphate
SMILES C1=CC=C([C-]=C1)C2=CC=CC=N2.C1=CC=C([C-]=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.F[P-](F)(F)(F)(F)F.[Ir+3]

Specifications

Melting Point 250°C
Quantity 200 mg
Formula Weight 801.75
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (2,2′-Bipyridine)bis(2-phenylpyridinato)iridium(III) Hexafluorophosphate