missing translation for 'onlineSavingsMsg'
Learn More

(2,2'-Bipyridine)bis[2-(2,4-difluorophenyl)pyridine]iridium(III) Hexafluorophosphate 80.0+%, TCI America™

Catalog No. b4944200mg
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4944200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4944200MG Supplier TCI America Supplier No. B4944200MG
Only null left

Chemical Identifiers

CAS 864163-80-4
Molecular Formula C32H20F10IrN4P
Molecular Weight (g/mol) 873.715
InChI Key BWQRXIXHHHVGJD-UHFFFAOYSA-N
PubChem CID 131637026
IUPAC Name 2-(2,4-difluorobenzene-6-id-1-yl)pyridine;iridium(3+);2-pyridin-2-ylpyridine;hexafluorophosphate
SMILES C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=C(C=C(C=[C-]2)F)F.C1=CC=NC(=C1)C2=C(C=C(C=[C-]2)F)F.F[P-](F)(F)(F)(F)F.[Ir+3]

Specifications

Quantity 200 mg
Formula Weight 873.71
Percent Purity ≥80.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (2,2′-Bipyridine)bis[2-(2,4-difluorophenyl)pyridine]iridium(III) Hexafluorophosphate